O-[(3-Pyridyl)methyl]hydroxylamine

Lieferant: Apollo Scientific

OR471001-5G
APOSOR471001-5GEA 1030 EUR
APOSOR471001-5G
O-[(3-Pyridyl)methyl]hydroxylamine
O-[(3-Pyridyl)methyl]hydroxylamine
Formel: C₆H₈N₂O
Molecular Weight: 124,141 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13175304
CAS-Nummer: 37832-20-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR