3-(Methoxymethoxy)phenylboronic acid pinacol ester

Lieferant: Apollo Scientific

OR471002-1G
APOSOR471002-1GEA 804 EUR
APOSOR471002-1G
3-(Methoxymethoxy)phenylboronic acid pinacol ester
3-(Methoxymethoxy)phenylboronic acid pinacol ester
Formel: C₁₄H₂₁BO₄
Molecular Weight: 264.125 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11856040
CAS-Nummer: 1245824-36-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR