3-Isopropyl-4-methoxybenzaldehyde

Lieferant: Apollo Scientific

OR471004-1G
APOSOR471004-1GEA 804 EUR
APOSOR471004-1G
3-Isopropyl-4-methoxybenzaldehyde
3-Isopropyl-4-methoxybenzaldehyde
Formel: C₁₁H₁₄O₂
Molecular Weight: 178.23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06247877
CAS-Nummer: 31825-29-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR