6-Methoxy[1,1'-biphenyl]-3-carbaldehyde
Lieferant: Apollo Scientific
OR471005-1G
APOSOR471005-1GEA
804
EUR
APOSOR471005-1G
6-Methoxy[1,1'-biphenyl]-3-carbaldehyde
6-Methoxy[1,1'-biphenyl]-3-carbaldehyde
Formel:
C₁₄H₁₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD06247960 CAS-Nummer: 258831-56-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...