Methyl 3'-Hydroxybiphenyl-4-carboxylate

Lieferant: Apollo Scientific

OR471006-1G OR471006-5G
APOSOR471006-1GEA 306 EUR
APOSOR471006-1G APOSOR471006-5G
Methyl 3'-Hydroxybiphenyl-4-carboxylate
Methyl 3'-Hydroxybiphenyl-4-carboxylate
Formel: C₁₄H₁₂O₃
Molecular Weight: 228.243 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06409247
CAS-Nummer: 579511-01-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR