3-(2-Bromo-4-nitrophenyl)propanoic acid

Lieferant: Apollo Scientific

OR471008-1G
APOSOR471008-1GEA 1010 EUR
APOSOR471008-1G
3-(2-Bromo-4-nitrophenyl)propanoic acid
3-(2-Bromo-4-nitrophenyl)propanoic acid
Formel: C₉H₈BrNO₄
Molecular Weight: 274,068 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12152254
CAS-Nummer: 1182426-40-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR