4,6-Nonandion

Lieferant: Apollo Scientific

OR471010-1G
APOSOR471010-1GEA 916 EUR
APOSOR471010-1G
4,6-Nonandion
4,6-Nonandion
Formel: C₉H₁₆O₂
Lagertemperatur: Raumtemperatur
CAS-Nummer: 14090-88-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR