Methyl 3-Amino-2-hydroxypropanoate hydrochloride

Lieferant: Apollo Scientific

OR471011-1G
APOSOR471011-1GEA 79.9 EUR
APOSOR471011-1G
Methyl 3-Amino-2-hydroxypropanoate hydrochloride
Methyl 3-Amino-2-hydroxypropanoate hydrochloride
Formel: C₄H₉NO₃·ClH
Molecular Weight: 155,58 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22741315
CAS-Nummer: 186393-00-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR