Methyl N-Boc-3-amino-2-hydroxypropanoate

Lieferant: Apollo Scientific

OR471013-1G
APOSOR471013-1GEA 1010 EUR
APOSOR471013-1G
Methyl N-Boc-3-amino-2-hydroxypropanoate
Methyl N-Boc-3-amino-2-hydroxypropanoate
Formel: C₉H₁₇NO₅
Molecular Weight: 219.235 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18447448
CAS-Nummer: 113525-87-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR