Methyl 2,4-Dimethyl-3-oxopentanoate

Lieferant: Apollo Scientific

OR471014-1G
APOSOR471014-1GEA 659 EUR
APOSOR471014-1G
Methyl 2,4-Dimethyl-3-oxopentanoate
Methyl 2,4-Dimethyl-3-oxopentanoate
Formel: C₈H₁₄O₃
Molecular Weight: 158.195 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09032945
CAS-Nummer: 59742-51-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR