1,2,2-Tris[(S)-4-isopropyl-4,5-dihydro-2-oxazolyl]propane
Lieferant: Apollo Scientific
|
Achtung
|
OR471017-1G
APOSOR471017-1GEA
1840
EUR
APOSOR471017-1G
1,2,2-Tris[(S)-4-isopropyl-4,5-dihydro-2-oxazolyl]propane
1,2,2-Tris[(S)-4-isopropyl-4,5-dihydro-2-oxazolyl]propane
Formel:
C₂₁H₃₅N₃O₃ Molecular Weight: 377,521 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD26404517 CAS-Nummer: 458563-75-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...