4-Bromo-3-chlorobenzyl Alcohol

Lieferant: Apollo Scientific

Synonyme: (4-Bromo-3-Chlorophenyl)methanol, 4-Bromo-3-chlorobenzyl alcohol

OR471021-5G
APOSOR471021-5GEA 499 EUR
APOSOR471021-5G
4-Bromo-3-chlorobenzyl Alcohol
4-Bromo-3-chlorobenzyl Alcohol
Formel: C₇H₆BrClO
Molecular Weight: 221,479 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08059111
CAS-Nummer: 120077-68-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR