4-Amino-1-methylpyrazole-3-carboxylic acid

Lieferant: Apollo Scientific

OR471022-1G
APOSOR471022-1GEA 374 EUR
APOSOR471022-1G
4-Amino-1-methylpyrazole-3-carboxylic acid
4-Amino-1-methylpyrazole-3-carboxylic acid
Formel: C₅H₇N₃O₂
Molecular Weight: 141,128 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08558492
CAS-Nummer: 1006334-34-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR