1-(Hydroxymethyl)indan

Lieferant: Apollo Scientific

OR471023-1G
APOSOR471023-1GEA 754 EUR
APOSOR471023-1G
1-(Hydroxymethyl)indan
1-(Hydroxymethyl)indan
Formel: C₁₀H₁₂O
Molecular Weight: 148,202 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18089511
CAS-Nummer: 1196-17-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR