3-Chlorotetrahydropyran-4-one

Lieferant: Apollo Scientific

OR471024-5G
APOSOR471024-5GEA 916 EUR
APOSOR471024-5G
3-Chlorotetrahydropyran-4-one
3-Chlorotetrahydropyran-4-one
Formel: C₅H₇ClO₂
Molecular Weight: 134,561 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11858310
CAS-Nummer: 160427-98-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR