Methyl 3-(3-Oxo-2-isothiazolyl)propanoate

Lieferant: Apollo Scientific

OR471025-1G
APOSOR471025-1GEA 1010 EUR
APOSOR471025-1G
Methyl 3-(3-Oxo-2-isothiazolyl)propanoate
Methyl 3-(3-Oxo-2-isothiazolyl)propanoate
Formel: C₇H₉NO₃S
Molecular Weight: 187,216 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD25962403
CAS-Nummer: 33319-78-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR