5-[Hydroxy(phenyl)methyl]dihydrofuran-2(3H)-one

Lieferant: Apollo Scientific

OR471026-1G
APOSOR471026-1GEA 1010 EUR
APOSOR471026-1G
5-[Hydroxy(phenyl)methyl]dihydrofuran-2(3H)-one
5-[Hydroxy(phenyl)methyl]dihydrofuran-2(3H)-one
Formel: C₁₁H₁₂O₃
Molecular Weight: 192,211 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28053835
CAS-Nummer: 92017-03-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR