3-Hydroxy-2,2-dimethyl-1-indanone

Lieferant: Apollo Scientific

OR471029-1G
APOSOR471029-1GEA 707 EUR
APOSOR471029-1G
3-Hydroxy-2,2-dimethyl-1-indanone
3-Hydroxy-2,2-dimethyl-1-indanone
Formel: C₁₁H₁₂O₂
Molecular Weight: 176,212 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09907921
CAS-Nummer: 59269-93-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR