4-(Iodomethyl)piperidine hydrochloride

Lieferant: Apollo Scientific

OR471030-5G
APOSOR471030-5GEA 1120 EUR
APOSOR471030-5G
4-(Iodomethyl)piperidine hydrochloride
4-(Iodomethyl)piperidine hydrochloride
Formel: C₆H₁₂IN·ClH
Molecular Weight: 261,532 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21098334
CAS-Nummer: 1353976-20-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR