3-(Bromomethyl)piperidine hydrochloride

Lieferant: Apollo Scientific

OR471031-5G
APOSOR471031-5GEA 1120 EUR
APOSOR471031-5G
3-(Bromomethyl)piperidine hydrochloride
3-(Bromomethyl)piperidine hydrochloride
Formel: C₆H₁₂BrN·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD21098325
CAS-Nummer: 1353976-07-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR