1-(3-Chloropropyl)imidazole

Lieferant: Apollo Scientific

OR471035-1G
APOSOR471035-1GEA 601 EUR
APOSOR471035-1G
1-(3-Chloropropyl)imidazole
1-(3-Chloropropyl)imidazole
Formel: C₆H₉ClN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09728547
CAS-Nummer: 53710-78-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR