1-Cyclopentylimidazole
Lieferant: Apollo Scientific
OR471036-5G
APOSOR471036-5GEA
1330
EUR
APOSOR471036-5G
1-Cyclopentylimidazole
1-Cyclopentylimidazole
Formel:
C₈H₁₂N₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD18889431 CAS-Nummer: 71614-58-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...