(S)-4-Benzyl-2-(hydroxymethyl)morpholine

Lieferant: Apollo Scientific

OR471039-5G
APOSOR471039-5GEA 916 EUR
APOSOR471039-5G
(S)-4-Benzyl-2-(hydroxymethyl)morpholine
(S)-4-Benzyl-2-(hydroxymethyl)morpholine
Formel: C₁₂H₁₇NO₂
Molecular Weight: 207,269 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10698663
CAS-Nummer: 132073-82-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR