1-Cyclopropyl-1-(4-methoxyphenyl)methylamine

Lieferant: Apollo Scientific

OR471040-1G
APOSOR471040-1GEA 0 EUR
APOSOR471040-1G
1-Cyclopropyl-1-(4-methoxyphenyl)methylamine
1-Cyclopropyl-1-(4-methoxyphenyl)methylamine
Formel: C₁₁H₁₅NO
Lagertemperatur: Raumtemperatur
MDL: MFCD00044830
CAS-Nummer: 54398-65-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR