2-(Tetrahydrothiopyran-4-yl)ethylamine

Lieferant: Apollo Scientific

OR471042-1G
APOSOR471042-1GEA 916 EUR
APOSOR471042-1G
2-(Tetrahydrothiopyran-4-yl)ethylamine
2-(Tetrahydrothiopyran-4-yl)ethylamine
Formel: C₇H₁₅NS
Molecular Weight: 145,266 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18374674
CAS-Nummer: 1262410-73-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR