Methyl 3-hydroxy-4,4-dimethylpentanoate

Lieferant: Apollo Scientific

OR471046-1G
APOSOR471046-1GEA 397 EUR
APOSOR471046-1G
Methyl 3-hydroxy-4,4-dimethylpentanoate
Methyl 3-hydroxy-4,4-dimethylpentanoate
Formel: C₈H₁₆O₃
Molecular Weight: 160,211 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28101621
CAS-Nummer: 150943-32-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR