N-[3-(2,2-Diethoxyethoxy)benzyl]-2,2-diethoxyethylamine

Lieferant: Apollo Scientific

OR471047-1G
APOSOR471047-1GEA 713 EUR
APOSOR471047-1G
N-[3-(2,2-Diethoxyethoxy)benzyl]-2,2-diethoxyethylamine
N-[3-(2,2-Diethoxyethoxy)benzyl]-2,2-diethoxyethylamine
Formel: C₁₉H₃₃NO₅
Molecular Weight: 355,469 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28101620
CAS-Nummer: 2006277-32-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR