3-Amino-2-hydroxy-N,N-dimethylbenzamide hydrochloride

Lieferant: Apollo Scientific

OR471049-1G
APOSOR471049-1GEA 804 EUR
APOSOR471049-1G
3-Amino-2-hydroxy-N,N-dimethylbenzamide hydrochloride
3-Amino-2-hydroxy-N,N-dimethylbenzamide hydrochloride
Formel: C₉H₁₂N₂O₂·ClH
Molecular Weight: 216,665 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18909379
CAS-Nummer: 1000993-70-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR