1-Methyl-4-oxocyclohexanecarboxylic acid

Lieferant: Apollo Scientific

OR471053-1G
APOSOR471053-1GEA 0 EUR
APOSOR471053-1G
1-Methyl-4-oxocyclohexanecarboxylic acid
1-Methyl-4-oxocyclohexanecarboxylic acid
Formel: C₈H₁₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD18374767
CAS-Nummer: 24463-41-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR