Methyl 2-(1-Aminocyclohexyl)acetate

Lieferant: Apollo Scientific

OR471055-1G
APOSOR471055-1GEA 529 EUR
APOSOR471055-1G
Methyl 2-(1-Aminocyclohexyl)acetate
Methyl 2-(1-Aminocyclohexyl)acetate
Formel: C₉H₁₇NO₂
Molecular Weight: 171,237 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07784119
CAS-Nummer: 178242-64-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR