3-Bromo-5-chloro-4-methylbenzonitrile
Lieferant: Apollo Scientific
OR471057-1G
APOSOR471057-1GEA
1220
EUR
APOSOR471057-1G
3-Bromo-5-chloro-4-methylbenzonitrile
3-Bromo-5-chloro-4-methylbenzonitrile
Formel:
C₈H₅BrClN Molecular Weight: 230.489 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11557540 CAS-Nummer: 939990-02-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...