Benzyl N-Boc-3-amino-5-iodopentanoate

Lieferant: Apollo Scientific

OR471058-250MG
APOSOR471058-250MGEA 1120 EUR
APOSOR471058-250MG
Benzyl N-Boc-3-amino-5-iodopentanoate
Benzyl N-Boc-3-amino-5-iodopentanoate
Formel: C₁₇H₂₄INO₄
Molecular Weight: 433,281 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28101613
CAS-Nummer: 643044-91-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR