3-Amino-N-(tert-butyl)propanamide hydrochloride
Lieferant: Apollo Scientific
Synonyme:
3-Amino-N-(tert-butyl)propanamide
OR471059-5G
APOSOR471059-5GEA
1120
EUR
APOSOR471059-5G
3-Amino-N-(tert-butyl)propanamide hydrochloride
3-Amino-N-(tert-butyl)propanamide hydrochloride
Formel:
C₇H₁₇ClN₂O Molecular Weight: 180,68 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11109909 CAS-Nummer: 209467-48-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...