2-(Chloromethyl)thiazole hydrochloride

Lieferant: Apollo Scientific

Synonyme: 2-(Chloromethyl)-1,3-thiazole hydrochloride

OR471060-1G
APOSOR471060-1GEA 591 EUR
APOSOR471060-1G
2-(Chloromethyl)thiazole hydrochloride
2-(Chloromethyl)thiazole hydrochloride
Formel: C₄H₄ClNS·ClH
Molecular Weight: 170,06 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09907885
CAS-Nummer: 71670-79-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR