Ethyl 1-Ethyl-2,4-dioxopiperidine-3-carboxylate

Lieferant: Apollo Scientific

OR471061-1G
APOSOR471061-1GEA 815 EUR
APOSOR471061-1G
Ethyl 1-Ethyl-2,4-dioxopiperidine-3-carboxylate
Ethyl 1-Ethyl-2,4-dioxopiperidine-3-carboxylate
Formel: C₁₀H₁₅NO₄
Molecular Weight: 213,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28122817
CAS-Nummer: 104966-05-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR