1-Bromo-3-hydroxynaphthalene
Lieferant: Apollo Scientific
|
Gefahr
|
OR471062-5G
APOSOR471062-5GEA
71.2
EUR
APOSOR471062-5G
1-Bromo-3-hydroxynaphthalene
1-Bromo-3-hydroxynaphthalene
Formel:
C₁₀H₇BrO Molecular Weight: 223.066 g/mol Siedepunkt: 130…132 °C (1 mm Hg) Schmelzpunkt: 117…120 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01712260 CAS-Nummer: 5498-31-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...