3-Cyclopropyl-3-(3-hydroxyphenyl)propanoic acid

Lieferant: Apollo Scientific

OR471063-5G
APOSOR471063-5GEA 1540 EUR
APOSOR471063-5G
3-Cyclopropyl-3-(3-hydroxyphenyl)propanoic acid
3-Cyclopropyl-3-(3-hydroxyphenyl)propanoic acid
Formel: C₁₂H₁₄O₃
Molecular Weight: 206,238 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24503676
CAS-Nummer: 1142224-60-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR