Methyl 6-Methoxy-2,3-dihydrobenzofuran-2-carboxylate

Lieferant: Apollo Scientific

OR471064-1G
APOSOR471064-1GEA 916 EUR
APOSOR471064-1G
Methyl 6-Methoxy-2,3-dihydrobenzofuran-2-carboxylate
Methyl 6-Methoxy-2,3-dihydrobenzofuran-2-carboxylate
Formel: C₁₁H₁₂O₄
Molecular Weight: 208,211 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD25961436
CAS-Nummer: 24007-57-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR