Methyl 1-(Methylsulphonyl)pyrrolidine-2-carboxylate

Lieferant: Apollo Scientific

OR471066-5G
APOSOR471066-5GEA 916 EUR
APOSOR471066-5G
Methyl 1-(Methylsulphonyl)pyrrolidine-2-carboxylate
Methyl 1-(Methylsulphonyl)pyrrolidine-2-carboxylate
Formel: C₇H₁₃NO₄S
Molecular Weight: 207,247 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08256096
CAS-Nummer: 1487500-62-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR