N-Boc-6-hydroxy-DL-norleucine methyl ester

Lieferant: Apollo Scientific

OR471067-1G OR471067-250MG
APOSOR471067-1GEA 1840 EUR
APOSOR471067-1G APOSOR471067-250MG
N-Boc-6-hydroxy-DL-norleucine methyl ester
N-Boc-6-hydroxy-DL-norleucine methyl ester
Formel: C₁₂H₂₃NO₅
Lagertemperatur: Raumtemperatur
MDL: MFCD18384776
CAS-Nummer: 81505-49-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR