2,6-Dihydroxyhexanenitrile
Lieferant: Apollo Scientific
OR471070-5G
APOSOR471070-5GEA
916
EUR
APOSOR471070-5G
2,6-Dihydroxyhexanenitrile
2,6-Dihydroxyhexanenitrile
Formel:
C₆H₁₁NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD19300782 CAS-Nummer: 17264-36-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...