Benzyl 2-Ethoxy-2-methylpropanoate

Lieferant: Apollo Scientific

OR471073-5G
APOSOR471073-5GEA 1120 EUR
APOSOR471073-5G
Benzyl 2-Ethoxy-2-methylpropanoate
Benzyl 2-Ethoxy-2-methylpropanoate
Formel: C₁₃H₁₈O₃
Molecular Weight: 222,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28122815
CAS-Nummer: 1000296-72-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR