4-Cbz-1-methyl-2-piperazinone

Lieferant: Apollo Scientific

OR471075-5G
APOSOR471075-5GEA 713 EUR
APOSOR471075-5G
4-Cbz-1-methyl-2-piperazinone
4-Cbz-1-methyl-2-piperazinone
Formel: C₁₃H₁₆N₂O₃
Molecular Weight: 248,278 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28122814
CAS-Nummer: 685520-31-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR