2-Amino-5-bromo-4,6-dimethyl-3-nitropyridine

Lieferant: Apollo Scientific

OR471076-1G
APOSOR471076-1GEA 561 EUR
APOSOR471076-1G
2-Amino-5-bromo-4,6-dimethyl-3-nitropyridine
2-Amino-5-bromo-4,6-dimethyl-3-nitropyridine
Formel: C₇H₈BrN₃O₂
Molecular Weight: 246,061 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03931432
CAS-Nummer: 89791-76-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR