Methyl 1-Cbz-2-methylpyrrolidine-2-carboxylate

Lieferant: Apollo Scientific

OR471077-1G
APOSOR471077-1GEA 448 EUR
APOSOR471077-1G
Methyl 1-Cbz-2-methylpyrrolidine-2-carboxylate
Methyl 1-Cbz-2-methylpyrrolidine-2-carboxylate
Formel: C₁₅H₁₉NO₄
Molecular Weight: 277,316 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23701515
CAS-Nummer: 141870-95-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR