2-Bromo-6-hydroxy-4-methoxybenzaldehyde

Lieferant: Apollo Scientific

OR471078-5G OR471078-1G
APOSOR471078-5GEA 1120 EUR
APOSOR471078-5G APOSOR471078-1G
2-Bromo-6-hydroxy-4-methoxybenzaldehyde
2-Bromo-6-hydroxy-4-methoxybenzaldehyde
Formel: C₈H₇BrO₃
Molecular Weight: 231.043 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23705610
CAS-Nummer: 861668-41-9

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR