Ethyl 1-Benzyl-3-(Boc-amino)pyrrolidine-3-carboxylate
Lieferant: Apollo Scientific
OR471081-1G
APOSOR471081-1GEA
570
EUR
APOSOR471081-1G
Ethyl 1-Benzyl-3-(Boc-amino)pyrrolidine-3-carboxylate
Ethyl 1-Benzyl-3-(Boc-amino)pyrrolidine-3-carboxylate
Formel:
C₁₉H₂₈N₂O₄ Molecular Weight: 348,437 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28122813 CAS-Nummer: 1823389-67-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...