1-Nitro-2-propoxybenzene

Lieferant: Apollo Scientific

OR471084-5G
APOSOR471084-5GEA 713 EUR
APOSOR471084-5G
1-Nitro-2-propoxybenzene
1-Nitro-2-propoxybenzene
Formel: C₉H₁₁NO₃
Molecular Weight: 181,189 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28122812
CAS-Nummer: 3079-53-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR