Methyl 2-(2-Bromo-5-methyl-4-thiazolyl)acetate

Lieferant: Apollo Scientific

Synonyme: Methyl 2-(2-bromo-5-methylthiazol-4-yl)acetate

OR471085-1G
APOSOR471085-1GEA 713 EUR
APOSOR471085-1G
Methyl 2-(2-Bromo-5-methyl-4-thiazolyl)acetate
Methyl 2-(2-Bromo-5-methyl-4-thiazolyl)acetate
Formel: C₇H₈BrNO₂S
Molecular Weight: 250,113 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09038182
CAS-Nummer: 496062-15-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR