N-[3-(Dimethylamino)propyl]acetamide

Lieferant: Apollo Scientific

OR471086-5G
APOSOR471086-5GEA 815 EUR
APOSOR471086-5G
N-[3-(Dimethylamino)propyl]acetamide
N-[3-(Dimethylamino)propyl]acetamide
Formel: C₇H₁₆N₂O
Molecular Weight: 144.215 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01195313
CAS-Nummer: 3197-19-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR